Gene Transcripts

Gene Symbol
Gbgt1
Entrez Gene ID
227671
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
Fgs, Fs
Chromosome Location
Chr. 2: 28479229 - 28506030 on Build GRCm38
UniGene ID
Mm.484270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139197.2 NP_631936.2 7 702
GenBank mRNA AB084789.1 - 6 451
AF292399.1 - 6 431
BC106831.2 - 6 431 40044790

View Full Product Details