Gene Transcripts
- Gene Symbol
- F9
- Entrez Gene ID
- 24946
- Gene Name
- coagulation factor IX
- Gene Aliases
- Chromosome Location
- Chr. X: 143097560 - 143141786 on Build Rnor_6.0
- UniGene ID
- Rn.37599
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031540.1 |
NP_113728.1
|
8 |
1177 |
|
GenBank mRNA |
M26247.1 |
- |
3 |
661 |
|
View Full Product Details