Gene Transcripts

Gene Symbol
Olr1253
Entrez Gene ID
405091
Gene Name
olfactory receptor 1253
Gene Aliases
Chromosome Location
Chr. 8: 42376325 - 42377257 on Build Rnor_6.0
UniGene ID
Rn.143517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000805.1 NP_001000805.1 1 577

Gene Symbol
LOC100912543
Entrez Gene ID
100912543
Gene Name
olfactory receptor 8B3-like
Gene Aliases
Chromosome Location
Chr. 3: 158931300 - 158932232 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762453.1 XP_008760675.1 1 577

Gene Symbol
LOC103692041
Entrez Gene ID
103692041
Gene Name
olfactory receptor 8B3-like
Gene Aliases
Chromosome Location
Chr. 4: 1697351 - 1698283 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762675.1 XP_008760897.1 1 577

View Full Product Details