Gene Transcripts

Gene Symbol
POMZP3
Entrez Gene ID
22932
Gene Name
POM121 and ZP3 fusion
Gene Aliases
POM-ZP3, POM121
Chromosome Location
Chr. 7: 76609986 - 76627303 on Build GRCh38
UniGene ID
Hs.488877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012230.3 NP_036362.3 2 613
NM_152992.2 NP_694537.1 2 613
GenBank mRNA U10099.1 - 2 559

Gene Symbol
POM121C
Entrez Gene ID
100101267
Gene Name
POM121 transmembrane nucleoporin C
Gene Aliases
POM121-2
Chromosome Location
Chr. 7: 75416781 - 75486288 on Build GRCh38
UniGene ID
Hs.712563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099415.2 NP_001092885.2 4 748
GenBank mRNA AB354586.1 - 2 592
BC082993.1 - 4 731
BC130587.1 - 4 455
BC130589.1 - 4 455

View Full Product Details