Gene Transcripts

Gene Symbol
KRTAP6-1
Entrez Gene ID
337966
Gene Name
keratin associated protein 6-1
Gene Aliases
C21orf103, KAP6.1
Chromosome Location
Chr. 21: 30613686 - 30613904 on Build GRCh38
UniGene ID
Hs.194301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181602.1 NP_853633.1 1 173
GenBank mRNA AB096954.1 - 1 199

View Full Product Details