Gene Transcripts

Gene Symbol
TNFRSF25
Entrez Gene ID
8718
Gene Name
TNF receptor superfamily member 25
Gene Aliases
APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
Chromosome Location
Chr. 1: 6461151 - 6466195 on Build GRCh38
UniGene ID
Hs.462529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039664.1 NP_001034753.1 3 342
NM_003790.2 NP_003781.1 3 342
NM_148965.1 NP_683866.1 3 342
NM_148966.1 NP_683867.1 3 342
GenBank mRNA AB308321.1 - 3 254
AF026070.1 - 3 322
AF026071.1 - 3 322
AY358309.1 - 3 356
BC117189.1 - 3 322 40125740
BC143886.1 - 3 322 9052402
U72763.1 - 3 254
U74611.1 - 3 342
U75380.1 - 3 245
U75381.1 - 3 245
U78029.1 - 3 254
U83597.1 - 3 238
U83598.1 - 3 238
U94501.1 - 3 254
U94502.1 - 3 254
U94503.1 - 3 254
U94504.1 - 3 254
U94505.1 - 3 254
U94510.1 - 3 254
U94512.1 - 3 246
Y09392.1 - 3 322

View Full Product Details