Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015227.4 | NP_056042.1 | 5 | 684 | |
NM_133635.4 | NP_598368.2 | 5 | 684 | ||
NR_004858.1 | 5 | 684 | |||
XM_006723981.3 | XP_006724044.1 | 6 | 757 | ||
XM_011529504.2 | XP_011527806.1 | 7 | 893 | ||
XM_011529505.2 | XP_011527807.1 | 7 | 894 | ||
XM_011529506.2 | XP_011527808.1 | 6 | 832 | ||
XM_011529507.2 | XP_011527809.1 | 6 | 728 | ||
XM_011529508.2 | XP_011527810.1 | 7 | 894 | ||
XM_011529509.2 | XP_011527811.1 | 7 | 895 | ||
XM_011529510.2 | XP_011527812.1 | 7 | 802 | ||
XM_011529511.2 | XP_011527813.1 | 7 | 834 | ||
XM_017028306.1 | XP_016883795.1 | 5 | 667 | ||
XM_017028309.1 | XP_016883798.1 | 3 | 90 | ||
GenBank mRNA | AB023175.1 | - | 5 | 673 | |
AJ302079.2 | - | 5 | 673 | ||
AJ302080.2 | - | 5 | 673 | ||
AJ575591.1 | - | 5 | 668 | ||
AK290732.1 | - | 5 | 681 | ||
AL110285.1 | - | 5 | 670 | ||
AY066015.2 | - | 5 | 673 | ||
BC064623.1 | - | 5 | 666 | 5727996 |