Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142364.1 | NP_001135836.1 | 3 | 327 | |
NM_013338.4 | NP_037470.1 | 3 | 327 | ||
GenBank mRNA | AF102850.1 | - | 3 | 335 | |
AF161498.1 | - | 3 | 276 | ||
AF183413.1 | - | 3 | 286 | ||
AK299085.1 | - | 3 | 303 | ||
AK299126.1 | - | 3 | 296 | ||
BC012531.1 | - | 3 | 279 | 4273297 | |
BF668393.1 | - | 301 | |||
BG482604.1 | - | 3 | 312 | ||
BP246765.1 | - | 3 | 327 | ||
CB988638.1 | - | 324 |