Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033087.3 | NP_149078.1 | 2 | 467 | |
NR_024532.1 | 3 | 626 | |||
GenBank mRNA | AB161356.1 | - | 2 | 424 | |
AK027417.1 | - | 2 | 419 | ||
AK027814.1 | - | 1 | 1219 | ||
AK074704.1 | - | 2 | 419 | ||
AK074988.1 | - | 3 | 604 | ||
AK075172.1 | - | 2 | 443 | ||
AY358697.1 | - | 2 | 211 | ||
BC017876.1 | - | 2 | 417 | 4698763 | |
DA076693.1 | - | 2 | 468 | ||
DB019023.1 | - | 2 | 469 |