Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr. 14: 88824153 - 88877996 on Build GRCh38
UniGene ID
Hs.303055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288781.1 NP_001275710.1 5 579
NM_001288782.1 NP_001275711.1 5 579
NM_001288783.1 NP_001275712.1 5 579
NM_144596.3 NP_653197.2 6 609
NM_198309.3 NP_938051.1 5 579
NM_198310.3 NP_938052.1 5 579
XM_006720035.1 XP_006720098.1 4 861
XM_006720037.2 XP_006720100.1 4 861
XM_011536432.1 XP_011534734.1 4 861
XM_011536433.1 XP_011534735.1 4 861
XM_011536434.1 XP_011534736.1 4 861
XM_011536435.2 XP_011534737.1 3 265
XM_017020977.1 XP_016876466.1 4 861
XM_017020978.1 XP_016876467.1 4 861
GenBank mRNA AK093891.1 - 4 457
AK124675.1 - 5 459
AK296077.1 - 4 440
AK315772.1 - 4 464
AL833901.1 - 4 403
AY366523.1 - 4 435
AY366524.1 - 4 435
AY373972.1 - 4 435
BC026351.1 - 4 435 4799330
BC095433.1 - 4 417 30368216
BG330023.1 - 456
BX161472.1 - 4 419
BX247959.1 - 6 733
BX248248.1 - 2 111
KU178849.1 - 4 383
KU178850.1 - 4 383
KU178851.1 - 4 383
KU178853.1 - 4 383

View Full Product Details