Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009179.3 | NP_033205.2 | 4, 5 | 1903 | |
GenBank mRNA | AK029246.1 | - | 4, 5 | 1818 | |
AK031052.1 | - | 4, 5 | 1846 | ||
AK034863.1 | - | 4, 5 | 1844 | ||
AK053827.1 | - | 4, 5 | 1842 | ||
AK131653.1 | - | 3, 4 | 726 | ||
BC015264.1 | - | 3, 4 | 1145 | 4211307 | |
BC066064.1 | - | 4, 5 | 1802 | 6850583 | |
X76989.1 | - | 3, 4 | 809 |