Gene Transcripts

Gene Symbol
Pgm1
Entrez Gene ID
66681
Gene Name
phosphoglucomutase 1
Gene Aliases
3230402E02Rik, Pgm-1, Pgm2
Chromosome Location
Chr. 5: 64092831 - 64128355 on Build GRCm38
UniGene ID
Mm.2325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025700.2 NP_079976.1 9, 10 1240
GenBank mRNA AK014332.1 - 9, 10 1165
BC030869.1 - 9, 10 1202
BC052762.1 - 9, 10 1240 6510389

View Full Product Details