Gene Transcripts

Gene Symbol
SLC12A7
Entrez Gene ID
10723
Gene Name
solute carrier family 12 member 7
Gene Aliases
KCC4
Chromosome Location
Chr. 5: 1050374 - 1155887 on Build GRCh38
UniGene ID
Hs.172613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006598.2 NP_006589.2 24 4683
XM_005248231.3 XP_005248288.1 25 4703
XM_011513939.2 XP_011512241.1 25 4792
XM_011513940.2 XP_011512242.1 25 4627
XM_011513941.2 XP_011512243.2 25 4722
XM_017008958.1 XP_016864447.1 25 5002
XM_017008959.1 XP_016864448.1 25 4654
GenBank mRNA AF105365.1 - 24 4621
AK024493.1 - 17 3708
AK024497.1 - 17 3533
AK026625.1 - 1 1354
AL117433.1 - 5 1927
BC007760.2 - 8 2059
BC018982.2 - 8 2059
BC098390.1 - 24 4661 5265743

View Full Product Details