Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253849.1 | NP_001240778.1 | 6 | 2641 | |
NM_001253850.1 | NP_001240779.1 | 5 | 2211 | ||
NM_024626.3 | NP_078902.2 | 6 | 2559 | ||
NR_045603.1 | 6 | 2649 | |||
NR_045604.1 | 6 | 2353 | |||
XM_011542143.2 | XP_011540445.2 | 6 | 2843 | ||
XM_017002335.1 | XP_016857824.1 | 7 | 2869 | ||
GenBank mRNA | AK026071.1 | - | 7 | 1721 | |
BX648021.1 | - | 7 | 2543 |