Gene Transcripts
- Gene Symbol
- ZNF276
- Entrez Gene ID
- 92822
- Gene Name
- zinc finger protein 276
- Gene Aliases
- CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
- Chromosome Location
- Chr. 16: 89720368 - 89740925 on Build GRCh38
- UniGene ID
- Hs.290154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113525.1 |
NP_001106997.1
|
7, 8 |
1371 |
|
NM_152287.3 |
NP_689500.2
|
7, 8 |
1361 |
|
NR_110122.1 |
|
7, 8 |
1361 |
|
NR_110126.1 |
|
7, 8 |
1329 |
|
NR_110128.1 |
|
7, 8 |
1252 |
|
NR_110129.1 |
|
7, 8 |
1341 |
|
XM_005256324.3 |
XP_005256381.1
|
7, 8 |
1320 |
|
XM_005256328.3 |
XP_005256385.1
|
5, 6 |
1001 |
|
XM_017023890.1 |
XP_016879379.1
|
6, 7 |
1296 |
|
GenBank mRNA |
AK026482.1 |
- |
7, 8 |
1311 |
|
AK056405.1 |
- |
7, 8 |
1312 |
|
AK289801.1 |
- |
7, 8 |
1220 |
|
AK295770.1 |
- |
5, 6 |
1033 |
|
AK315793.1 |
- |
4, 5 |
728 |
|
BC032781.1 |
- |
6, 7 |
1030 |
|
BC105959.1 |
- |
7, 8 |
1154 |
40036181 |
BC107603.1 |
- |
7, 8 |
1186 |
40036180 |
BC110348.1 |
- |
7, 8 |
1086 |
5196957 |
View Full Product Details