Gene Transcripts
- Gene Symbol
- SETD4
- Entrez Gene ID
- 54093
- Gene Name
- SET domain containing 4
- Gene Aliases
- C21orf18, C21orf27
- Chromosome Location
- Chr. 21: 36034541 - 36060818 on Build GRCh38
- UniGene ID
- Hs.606200
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007259.2 |
NP_001007260.1
|
4, 5 |
383 |
|
NM_001007261.2 |
NP_001007262.1
|
5, 6 |
477 |
|
NM_001286752.1 |
NP_001273681.1
|
5, 6 |
866 |
|
NM_017438.4 |
NP_059134.1
|
4, 5 |
723 |
|
XM_011529636.1 |
XP_011527938.1
|
4, 5 |
772 |
|
XM_011529637.1 |
XP_011527939.1
|
4, 5 |
837 |
|
XM_011529638.2 |
XP_011527940.1
|
4, 5 |
745 |
|
XM_011529639.1 |
XP_011527941.1
|
4, 5 |
523 |
|
XM_011529640.2 |
XP_011527942.1
|
4, 5 |
788 |
|
XM_011529642.1 |
XP_011527944.1
|
5, 6 |
866 |
|
XM_011529643.1 |
XP_011527945.1
|
5, 6 |
817 |
|
XM_011529644.1 |
XP_011527946.1
|
5, 6 |
378 |
|
XM_017028403.1 |
XP_016883892.1
|
4, 5 |
314 |
|
XM_017028404.1 |
XP_016883893.1
|
5, 6 |
422 |
|
XM_017028405.1 |
XP_016883894.1
|
4, 5 |
723 |
|
GenBank mRNA |
AB004848.1 |
- |
5, 6 |
430 |
|
AB004852.1 |
- |
3, 4 |
254 |
|
AB004853.1 |
- |
5, 6 |
426 |
|
AK001660.1 |
- |
4, 5 |
290 |
|
AK095215.1 |
- |
3, 4 |
1590 |
|
BC002898.2 |
- |
4, 5 |
322 |
3940849 |
BC036556.2 |
- |
5, 6 |
540 |
5266627 |
BF195582.1 |
- |
4, 5 |
383 |
|
View Full Product Details