Gene Transcripts

Gene Symbol
VSTM4
Entrez Gene ID
196740
Gene Name
V-set and transmembrane domain containing 4
Gene Aliases
C10orf72
Chromosome Location
Chr. 10: 49014245 - 49129118 on Build GRCh38
UniGene ID
Hs.522928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144984.3 NP_659421.1 3 1435
GenBank mRNA AK056299.1 - 3 1412

View Full Product Details