Gene Transcripts

Gene Symbol
Setd1a
Entrez Gene ID
233904
Gene Name
SET domain containing 1A
Gene Aliases
BC010250, KMT2F, Nsccn1, mKIAA0339, mNSC1
Chromosome Location
Chr. 7: 127777389 - 127800122 on Build GRCm38
UniGene ID
Mm.435494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178029.3 NP_821172.2 19 5582
GenBank mRNA BC010250.1 - 6 1244
BC042890.1 - 6 1670
BC049883.1 - 12 2971

View Full Product Details