Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042659.1 | NP_001036124.1 | 2 | 940 | |
NM_022721.3 | NP_073558.2 | 2 | 1047 | ||
GenBank mRNA | AB052910.1 | - | 1 | 176 | |
AF272146.1 | - | 1 | 176 | ||
AK030111.1 | - | 2 | 1049 | ||
AK039402.1 | - | 2 | 939 | ||
BC117723.2 | - | 2 | 473 | 40088671 |