Gene Transcripts

Gene Symbol
Olfr1394
Entrez Gene ID
258273
Gene Name
olfactory receptor 1394
Gene Aliases
MOR280-1
Chromosome Location
Chr. 11: 49160016 - 49160954 on Build GRCm38
UniGene ID
Mm.377407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146276.1 NP_666388.1 1 589
GenBank mRNA BC127968.1 - 1 593

View Full Product Details