Gene Transcripts

Gene Symbol
Ugt3a1
Entrez Gene ID
105887
Gene Name
UDP glycosyltransferases 3 family, polypeptide A1
Gene Aliases
AI746432
Chromosome Location
Chr. 15: 9279826 - 9315038 on Build GRCm38
UniGene ID
Mm.482274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207216.2 NP_997099.2 3 337
GenBank mRNA AI746748.1 - 3 316
AK128903.1 - 3 309
AK143745.1 - 3 315
BC025940.1 - 3 304 5100753

View Full Product Details