Gene Transcripts

Gene Symbol
1700102P08Rik
Entrez Gene ID
112418
Gene Name
RIKEN cDNA 1700102P08 gene
Gene Aliases
AI593206
Chromosome Location
Chr. 9: 108392834 - 108410966 on Build GRCm38
UniGene ID
Mm.78287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053216.2 NP_444446.2 2 278
GenBank mRNA AK007115.1 - 2 278
BC061048.1 - 2 278 6770968

View Full Product Details