Gene Transcripts

Gene Symbol
Prdx3
Entrez Gene ID
64371
Gene Name
peroxiredoxin 3
Gene Aliases
Prx3
Chromosome Location
Chr. 1: 282238774 - 282251193 on Build Rnor_6.0
UniGene ID
Rn.2011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022540.1 NP_071985.1 2 149
GenBank mRNA AF106944.1 - 2 149
BC060567.1 - 2 150 6922338
FQ212826.1 - 2 129
FQ219655.1 - 2 146

View Full Product Details