Gene Transcripts

Gene Symbol
Slc25a10
Entrez Gene ID
170943
Gene Name
solute carrier family 25 (mitochondrial carrier; dicarboxylate transporter), member 10
Gene Aliases
Dic
Chromosome Location
Chr. 10: 109665682 - 109673143 on Build Rnor_6.0
UniGene ID
Rn.3631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133418.1 NP_596909.1 4 396
GenBank mRNA AJ223355.1 - 4 396
BC081734.1 - 4 414 7108552

View Full Product Details