Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172074.3 | NP_742071.1 | 2 | 729 | |
XM_006234473.2 | XP_006234535.1 | 3 | 341 | ||
XM_006234474.2 | XP_006234536.1 | 4 | 453 | ||
GenBank mRNA | AF150091.1 | - | 2 | 164 | |
BC091116.1 | - | 2 | 730 | 7366212 |