Gene Transcripts

Gene Symbol
Olr1107
Entrez Gene ID
300202
Gene Name
olfactory receptor 1107
Gene Aliases
Chromosome Location
Chr. 7: 140030438 - 140037711 on Build Rnor_6.0
UniGene ID
Rn.143286
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000424.1 NP_001000424.1 1 913
XM_006242320.2 XP_006242382.1 2 1019

View Full Product Details