Gene Transcripts

Gene Symbol
Olr1105
Entrez Gene ID
405968
Gene Name
olfactory receptor 1105
Gene Aliases
Chromosome Location
Chr. 7: 139953082 - 139953999 on Build Rnor_6.0
UniGene ID
Rn.143288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001078.1 NP_001001078.1 1 266

View Full Product Details