Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083614.1 | NP_001077083.1 | 8 | 1406 | |
NM_001308211.1 | NP_001295140.1 | 8 | 1406 | ||
NR_003501.1 | 8 | 1406 | |||
XM_011545738.1 | XP_011544040.1 | 8 | 1696 | ||
XM_011545739.1 | XP_011544041.1 | 8 | 1315 | ||
GenBank mRNA | AB075850.1 | - | 8 | 1363 |