Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130110.1 | NP_001123582.1 | 2 | 703 | |
NM_015559.2 | NP_056374.2 | 2 | 604 | ||
GenBank mRNA | AB007897.2 | - | 2 | 604 | |
AB022660.1 | - | 1 | 151 | ||
BC062338.1 | - | 2 | 703 | ||
BC146776.1 | - | 2 | 604 | 9007225 |