Gene Transcripts

Gene Symbol
VSIG10L
Entrez Gene ID
147645
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
Chromosome Location
Chr. 19: 51331541 - 51342124 on Build GRCh38
UniGene ID
Hs.293236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163922.1 NP_001157394.1 10 3104
GenBank mRNA BC016993.1 - 7 1723
BC032383.1 - 7 1717
BC049212.1 - 7 1770

View Full Product Details