Gene Transcripts
- Gene Symbol
- LOC107983983
- Entrez Gene ID
- 107983983
- Gene Name
- putative V-set and immunoglobulin domain-containing-like protein IGHV4OR15-8
- Gene Aliases
- Chromosome Location
- Chr. 15: 22183503 - 22185525 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- IGHD
- Entrez Gene ID
- 3495
- Gene Name
- immunoglobulin heavy constant delta
- Gene Aliases
- Chromosome Location
- Chr. 14: 105838401 - 105845677 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC063384.1 |
- |
2 |
172 |
|
- Gene Symbol
- LOC102724971
- Entrez Gene ID
- 102724971
- Gene Name
- putative V-set and immunoglobulin domain-containing-like protein IGHV4OR15-8
- Gene Aliases
- Chromosome Location
- Chr. 15: 21742069 - 21742916 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details