Gene Transcripts
- Gene Symbol
- LOC100133331
- Entrez Gene ID
- 100133331
- Gene Name
- uncharacterized LOC100133331
- Gene Aliases
- Chromosome Location
- Chr. 1: 725759 - 730351 on Build GRCh38
- UniGene ID
- Hs.728864
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028327.1 |
|
3 |
4059 |
|
GenBank mRNA |
AK097080.1 |
- |
4 |
2207 |
|
- Gene Symbol
- LINC00265
- Entrez Gene ID
- 349114
- Gene Name
- long intergenic non-protein coding RNA 265
- Gene Aliases
- NCRNA00265
- Chromosome Location
- Chr. 7: 39733568 - 39794623 on Build GRCh38
- UniGene ID
- Hs.414183
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026999.1 |
|
12 |
4552 |
|
GenBank mRNA |
AK097404.1 |
- |
2 |
2096 |
|
- Gene Symbol
- LINC00999
- Entrez Gene ID
- 399744
- Gene Name
- long intergenic non-protein coding RNA 999
- Gene Aliases
- Chromosome Location
- Chr. 10: 38428146 - 38452153 on Build GRCh38
- UniGene ID
- Hs.497951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024497.2 |
|
8 |
5039 |
|
GenBank mRNA |
AK128746.1 |
- |
2 |
3498 |
|
- Gene Symbol
- LINC01000
- Entrez Gene ID
- 402483
- Gene Name
- long intergenic non-protein coding RNA 1000
- Gene Aliases
- Chromosome Location
- Chr. 7: 128641241 - 128660998 on Build GRCh38
- UniGene ID
- Hs.722350
- Species
- Homo sapiens
- Gene Symbol
- LINC01001
- Entrez Gene ID
- 100133161
- Gene Name
- long intergenic non-protein coding RNA 1001
- Gene Aliases
- Chromosome Location
- Chr. 11: 126987 - 131920 on Build GRCh38
- UniGene ID
- Hs.645398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028326.1 |
|
3 |
4400 |
|
- Gene Symbol
- LINC01002
- Entrez Gene ID
- 399844
- Gene Name
- long intergenic non-protein coding RNA 1002
- Gene Aliases
- Chromosome Location
- Chr. 19: 197016 - 202209 on Build GRCh38
- UniGene ID
- Hs.356758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028324.1 |
|
3 |
4771 |
|
GenBank mRNA |
AK128780.1 |
- |
2 |
2076 |
|
BC117566.1 |
- |
1 |
822 |
|
- Gene Symbol
- LOC100132062
- Entrez Gene ID
- 100132062
- Gene Name
- uncharacterized LOC100132062
- Gene Aliases
- Chromosome Location
- Chr. 5: 181323506 - 181328195 on Build GRCh38
- UniGene ID
- Hs.722350
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028325.1 |
|
3 |
4156 |
|
GenBank mRNA |
BC112924.1 |
- |
2 |
2507 |
|
- Gene Symbol
- LOC100132217
- Entrez Gene ID
- 100132217
- Gene Name
- uncharacterized LOC100132217
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.520384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128836.1 |
- |
3 |
3941 |
|
- Gene Symbol
- LOC100132287
- Entrez Gene ID
- 100132287
- Gene Name
- uncharacterized LOC100132287
- Gene Aliases
- Chromosome Location
- Chr. 1: 490756 - 495445 on Build GRCh38
- UniGene ID
- Hs.446409
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028322.1 |
|
3 |
4156 |
|
- Gene Symbol
- LOC107986805
- Entrez Gene ID
- 107986805
- Gene Name
- putative uncharacterized protein FLJ44672
- Gene Aliases
- Chromosome Location
- Chr. 7: 63765043 - 63767827 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC401357
- Entrez Gene ID
- 401357
- Gene Name
- uncharacterized LOC401357
- Gene Aliases
- Chromosome Location
- Chr. 7: 56809124 - 56812736 on Build GRCh38
- UniGene ID
- Hs.457696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130727.1 |
|
1 |
3526 |
|
GenBank mRNA |
AK125790.1 |
- |
1 |
3525 |
|
AK128114.1 |
- |
1 |
3267 |
|
- Gene Symbol
- LOC729218
- Entrez Gene ID
- 729218
- Gene Name
- uncharacterized LOC729218
- Gene Aliases
- Chromosome Location
- Chr. 4: 118591744 - 118635025 on Build GRCh38
- UniGene ID
- Hs.448518
- Species
- Homo sapiens
- Gene Symbol
- LOC729737
- Entrez Gene ID
- 729737
- Gene Name
- uncharacterized LOC729737
- Gene Aliases
- Chromosome Location
- Chr. 1: 134773 - 140566 on Build GRCh38
- UniGene ID
- Hs.534942
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039983.2 |
|
3 |
5260 |
|
View Full Product Details