Gene Transcripts
- Gene Symbol
- RPL13A
- Entrez Gene ID
- 23521
- Gene Name
- ribosomal protein L13a
- Gene Aliases
- L13A, TSTA1
- Chromosome Location
- Chr. 19: 49487554 - 49492308 on Build GRCh38
- UniGene ID
- Hs.523185
- Species
- Homo sapiens
- Gene Symbol
- RPL13AP20
- Entrez Gene ID
- 387841
- Gene Name
- ribosomal protein L13a pseudogene 20
- Gene Aliases
- RPL13A_9_1211
- Chromosome Location
- Chr. 12: 12875477 - 12876136 on Build GRCh38
- UniGene ID
- Hs.631728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003932.2 |
|
1 |
95 |
|
- Gene Symbol
- RPL13AP5
- Entrez Gene ID
- 728658
- Gene Name
- ribosomal protein L13a pseudogene 5
- Gene Aliases
- RPL13A_8_1086, bA196N24.2
- Chromosome Location
- Chr. 10: 96750266 - 96750923 on Build GRCh38
- UniGene ID
- Hs.645344
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026712.1 |
|
1 |
95 |
|
- Gene Symbol
- RPL13AP6
- Entrez Gene ID
- 644511
- Gene Name
- ribosomal protein L13a pseudogene 6
- Gene Aliases
- RPL13A_7_1096, bA348N5.5
- Chromosome Location
- Chr. 10: 110936603 - 110937255 on Build GRCh38
- UniGene ID
- Hs.632083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026715.1 |
|
1 |
95 |
|
View Full Product Details