Gene Transcripts
- Gene Symbol
- RPL13AP20
- Entrez Gene ID
- 387841
- Gene Name
- ribosomal protein L13a pseudogene 20
- Gene Aliases
- RPL13A_9_1211
- Chromosome Location
- Chr. 12: 12875477 - 12876136 on Build GRCh38
- UniGene ID
- Hs.631728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003932.2 |
|
1 |
209 |
|
View Full Product Details