Gene Transcripts
- Gene Symbol
- CYP2G1P
- Entrez Gene ID
- 22952
- Gene Name
- cytochrome P450 family 2 subfamily G member 1, pseudogene
- Gene Aliases
- CYP2G1, CYP2G2P, CYP2GP1
- Chromosome Location
- Chr. 19: 40890826 - 40900508 on Build GRCh38
- UniGene ID
- Hs.738699
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040249.1 |
|
3 |
1757 |
|
GenBank mRNA |
AK127151.1 |
- |
3 |
1757 |
|
View Full Product Details