Gene Transcripts
- Gene Symbol
- TFCP2
- Entrez Gene ID
- 7024
- Gene Name
- transcription factor CP2
- Gene Aliases
- LBP1C, LSF, LSF1D, SEF, TFCP2C
- Chromosome Location
- Chr. 12: 51093756 - 51173143 on Build GRCh38
- UniGene ID
- Hs.48849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U03495.1 |
- |
1 |
12 |
|
View Full Product Details