Gene Transcripts
- Gene Symbol
- IGHM
- Entrez Gene ID
- 3507
- Gene Name
- immunoglobulin heavy constant mu
- Gene Aliases
- AGM1, MU, VH
- Chromosome Location
- Chr. 14: 105851966 - 105856217 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
- Gene Symbol
- ZCWPW2
- Entrez Gene ID
- 152098
- Gene Name
- zinc finger CW-type and PWWP domain containing 2
- Gene Aliases
- ZCW2
- Chromosome Location
- Chr. 3: 28349149 - 28526366 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB087877.1 |
- |
3 |
640 |
|
View Full Product Details