Gene Transcripts
- Gene Symbol
- IGHM
- Entrez Gene ID
- 3507
- Gene Name
- immunoglobulin heavy constant mu
- Gene Aliases
- AGM1, MU, VH
- Chromosome Location
- Chr. 14: 105851966 - 105856217 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
- Gene Symbol
- IGHV3OR16-7
- Entrez Gene ID
- 28309
- Gene Name
- immunoglobulin heavy variable 3/OR16-7 (pseudogene)
- Gene Aliases
- IGH, IGHV, IGHV3/OR16-7, IGHV3OR167
- Chromosome Location
- Chr. 16: 33938339 - 33938639 on Build GRCh38
- UniGene ID
- Hs.649847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF209733.1 |
- |
3 |
378 |
|
- Gene Symbol
- LOC102723407
- Entrez Gene ID
- 102723407
- Gene Name
- putative V-set and immunoglobulin domain-containing-like protein IGHV4OR15-8
- Gene Aliases
- IGH, IGH@, IGHV4, IGHV4-30, IGHV4-b, IGVH, IGVH4-39, IGVH4-b, IGVH4b, IgA, IgT, ighv
- Chromosome Location
- Chr. 14|NT_187381: 41382 - 42173 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
- Gene Symbol
- ZCWPW2
- Entrez Gene ID
- 152098
- Gene Name
- zinc finger CW-type and PWWP domain containing 2
- Gene Aliases
- ZCW2
- Chromosome Location
- Chr. 3: 28349149 - 28526366 on Build GRCh38
- UniGene ID
- Hs.510635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB087877.1 |
- |
3 |
456 |
|
View Full Product Details