Gene Transcripts
- Gene Symbol
- SETD4
- Entrez Gene ID
- 54093
- Gene Name
- SET domain containing 4
- Gene Aliases
- C21orf18, C21orf27
- Chromosome Location
- Chr. 21: 36034541 - 36060818 on Build GRCh38
- UniGene ID
- Hs.606200
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007259.2 |
NP_001007260.1
|
2, 3 |
246 |
|
NM_017438.4 |
NP_059134.1
|
2, 3 |
586 |
|
NR_040087.2 |
|
2, 3 |
586 |
|
XM_011529636.1 |
XP_011527938.1
|
2, 3 |
635 |
|
XM_011529637.1 |
XP_011527939.1
|
2, 3 |
700 |
|
XM_011529638.2 |
XP_011527940.1
|
2, 3 |
608 |
|
XM_011529639.1 |
XP_011527941.1
|
2, 3 |
386 |
|
XM_011529640.2 |
XP_011527942.1
|
2, 3 |
651 |
|
XM_017028403.1 |
XP_016883892.1
|
2, 3 |
177 |
|
XM_017028405.1 |
XP_016883894.1
|
2, 3 |
586 |
|
GenBank mRNA |
AB004852.1 |
- |
1, 2 |
117 |
|
AF391112.1 |
- |
2, 3 |
153 |
|
AK001660.1 |
- |
2, 3 |
153 |
|
AK095215.1 |
- |
1, 2 |
1453 |
|
AK300009.1 |
- |
2, 3 |
286 |
|
BC002898.2 |
- |
2, 3 |
185 |
3940849 |
BF195582.1 |
- |
2, 3 |
246 |
|
View Full Product Details