Gene Transcripts
- Gene Symbol
- ZFP91
- Entrez Gene ID
- 80829
- Gene Name
- ZFP91 zinc finger protein
- Gene Aliases
- DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
- Chromosome Location
- Chr. 11: 58579114 - 58621550 on Build GRCh38
- UniGene ID
- Hs.524920
- Species
- Homo sapiens
- Gene Symbol
- ZFP91-CNTF
- Entrez Gene ID
- 386607
- Gene Name
- ZFP91-CNTF readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 11: 58579114 - 58625733 on Build GRCh38
- UniGene ID
- Hs.524920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024091.1 |
|
11 |
1474 |
|
GenBank mRNA |
AB057443.1 |
- |
11 |
1420 |
|
View Full Product Details