Gene Transcripts
- Gene Symbol
- LINC01000
- Entrez Gene ID
- 402483
- Gene Name
- long intergenic non-protein coding RNA 1000
- Gene Aliases
- Chromosome Location
- Chr. 7: 128641241 - 128660998 on Build GRCh38
- UniGene ID
- Hs.722350
- Species
- Homo sapiens
- Gene Symbol
- GTF2IP20
- Entrez Gene ID
- 441124
- Gene Name
- general transcription factor IIi pseudogene 20
- Gene Aliases
- Chromosome Location
- Chr. 1: 223951266 - 223992644 on Build GRCh38
- UniGene ID
- Hs.520589
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132119.1 |
|
11 |
2401 |
|
GenBank mRNA |
AK123446.1 |
- |
1 |
1628 |
|
BQ932773.1 |
- |
6 |
824 |
|
- Gene Symbol
- LINC00265
- Entrez Gene ID
- 349114
- Gene Name
- long intergenic non-protein coding RNA 265
- Gene Aliases
- NCRNA00265
- Chromosome Location
- Chr. 7: 39733568 - 39794623 on Build GRCh38
- UniGene ID
- Hs.414183
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026999.1 |
|
12 |
2240 |
|
GenBank mRNA |
AK097404.1 |
- |
1 |
319 |
|
- Gene Symbol
- LINC00999
- Entrez Gene ID
- 399744
- Gene Name
- long intergenic non-protein coding RNA 999
- Gene Aliases
- Chromosome Location
- Chr. 10: 38428146 - 38452153 on Build GRCh38
- UniGene ID
- Hs.497951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024497.2 |
|
8 |
4703 |
|
GenBank mRNA |
AK128746.1 |
- |
2 |
3162 |
|
- Gene Symbol
- LINC01001
- Entrez Gene ID
- 100133161
- Gene Name
- long intergenic non-protein coding RNA 1001
- Gene Aliases
- Chromosome Location
- Chr. 11: 126987 - 131920 on Build GRCh38
- UniGene ID
- Hs.645398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028326.1 |
|
3 |
1607 |
|
- Gene Symbol
- LINC01002
- Entrez Gene ID
- 399844
- Gene Name
- long intergenic non-protein coding RNA 1002
- Gene Aliases
- Chromosome Location
- Chr. 19: 197016 - 202209 on Build GRCh38
- UniGene ID
- Hs.356758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028324.1 |
|
3 |
4407 |
|
GenBank mRNA |
AK128780.1 |
- |
2 |
1712 |
|
BC117566.1 |
- |
1 |
458 |
|
- Gene Symbol
- LINC01061
- Entrez Gene ID
- 401149
- Gene Name
- long intergenic non-protein coding RNA 1061
- Gene Aliases
- Chromosome Location
- Chr. 4: 119405523 - 119410660 on Build GRCh38
- UniGene ID
- Hs.448518
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037596.1 |
|
3 |
3444 |
|
- Gene Symbol
- LOC100132062
- Entrez Gene ID
- 100132062
- Gene Name
- uncharacterized LOC100132062
- Gene Aliases
- Chromosome Location
- Chr. 5: 181323506 - 181328195 on Build GRCh38
- UniGene ID
- Hs.722350
- Species
- Homo sapiens
- Gene Symbol
- LOC100132287
- Entrez Gene ID
- 100132287
- Gene Name
- uncharacterized LOC100132287
- Gene Aliases
- Chromosome Location
- Chr. 1: 490756 - 495445 on Build GRCh38
- UniGene ID
- Hs.446409
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028322.1 |
|
3 |
1607 |
|
GenBank mRNA |
AK311126.1 |
- |
1 |
781 |
|
- Gene Symbol
- LOC100133331
- Entrez Gene ID
- 100133331
- Gene Name
- uncharacterized LOC100133331
- Gene Aliases
- Chromosome Location
- Chr. 1: 725759 - 730351 on Build GRCh38
- UniGene ID
- Hs.728864
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028327.1 |
|
3 |
1607 |
|
GenBank mRNA |
AK097080.1 |
- |
4 |
1843 |
|
- Gene Symbol
- LOC101060022
- Entrez Gene ID
- 101060022
- Gene Name
- nascent polypeptide-associated complex subunit alpha, muscle-specific form-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 227968754 - 227971943 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC105376666
- Entrez Gene ID
- 105376666
- Gene Name
- putative uncharacterized protein FLJ44672
- Gene Aliases
- Chromosome Location
- Chr. 11: 50070331 - 50076001 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC729218
- Entrez Gene ID
- 729218
- Gene Name
- uncharacterized LOC729218
- Gene Aliases
- Chromosome Location
- Chr. 4: 118591744 - 118635025 on Build GRCh38
- UniGene ID
- Hs.448518
- Species
- Homo sapiens
- Gene Symbol
- LOC729737
- Entrez Gene ID
- 729737
- Gene Name
- uncharacterized LOC729737
- Gene Aliases
- Chromosome Location
- Chr. 1: 134773 - 140566 on Build GRCh38
- UniGene ID
- Hs.534942
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_039983.2 |
|
3 |
1930 |
|
GenBank mRNA |
AK294489.1 |
- |
1 |
1263 |
|
AK302573.1 |
- |
1 |
893 |
|
View Full Product Details