Gene Transcripts
- Gene Symbol
- EFCAB2
- Entrez Gene ID
- 84288
- Gene Name
- EF-hand calcium binding domain 2
- Gene Aliases
- CFAP200, DRC8
- Chromosome Location
- Chr. 1: 244969739 - 245125228 on Build GRCh38
- UniGene ID
- Hs.134857
- Species
- Homo sapiens
- Gene Symbol
- ABCC5
- Entrez Gene ID
- 10057
- Gene Name
- ATP binding cassette subfamily C member 5
- Gene Aliases
- ABC33, EST277145, MOAT-C, MOATC, MRP5, SMRP, pABC11
- Chromosome Location
- Chr. 3: 183919934 - 184018010 on Build GRCh38
- UniGene ID
- Hs.368563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015334.1 |
- |
2 |
355 |
|
- Gene Symbol
- ALS2
- Entrez Gene ID
- 57679
- Gene Name
- ALS2, alsin Rho guanine nucleotide exchange factor
- Gene Aliases
- ALS2CR6, ALSJ, IAHSP, PLSJ
- Chromosome Location
- Chr. 2: 201700263 - 201781172 on Build GRCh38
- UniGene ID
- Hs.471096
- Species
- Homo sapiens
- Gene Symbol
- ANKRD28
- Entrez Gene ID
- 23243
- Gene Name
- ankyrin repeat domain 28
- Gene Aliases
- PITK, PPP1R65
- Chromosome Location
- Chr. 3: 15667236 - 15859814 on Build GRCh38
- UniGene ID
- Hs.335239
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126888.1 |
- |
20 |
3147 |
|
- Gene Symbol
- ARCN1
- Entrez Gene ID
- 372
- Gene Name
- archain 1
- Gene Aliases
- COPD
- Chromosome Location
- Chr. 11: 118572363 - 118603033 on Build GRCh38
- UniGene ID
- Hs.33642
- Species
- Homo sapiens
- Gene Symbol
- CASP2
- Entrez Gene ID
- 835
- Gene Name
- caspase 2
- Gene Aliases
- CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
- Chromosome Location
- Chr. 7: 143288215 - 143307696 on Build GRCh38
- UniGene ID
- Hs.368982
- Species
- Homo sapiens
- Gene Symbol
- CENPC
- Entrez Gene ID
- 1060
- Gene Name
- centromere protein C
- Gene Aliases
- CENP-C, CENPC1, MIF2, hcp-4
- Chromosome Location
- Chr. 4: 67468762 - 67545601 on Build GRCh38
- UniGene ID
- Hs.479867
- Species
- Homo sapiens
- Gene Symbol
- CHMP7
- Entrez Gene ID
- 91782
- Gene Name
- charged multivesicular body protein 7
- Gene Aliases
- Chromosome Location
- Chr. 8: 23243637 - 23262000 on Build GRCh38
- UniGene ID
- Hs.5019
- Species
- Homo sapiens
- Gene Symbol
- DENR
- Entrez Gene ID
- 8562
- Gene Name
- density regulated re-initiation and release factor
- Gene Aliases
- DRP, DRP1, SMAP-3
- Chromosome Location
- Chr. 12: 122752821 - 122771064 on Build GRCh38
- UniGene ID
- Hs.22393
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF038554.1 |
- |
8 |
1460 |
|
- Gene Symbol
- DHRS1
- Entrez Gene ID
- 115817
- Gene Name
- dehydrogenase/reductase 1
- Gene Aliases
- SDR19C1
- Chromosome Location
- Chr. 14: 24290598 - 24299833 on Build GRCh38
- UniGene ID
- Hs.348350
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126383.1 |
- |
1 |
311 |
|
- Gene Symbol
- EWSR1
- Entrez Gene ID
- 2130
- Gene Name
- EWS RNA binding protein 1
- Gene Aliases
- EWS, EWS-FLI1, bK984G1.4
- Chromosome Location
- Chr. 22: 29268009 - 29300525 on Build GRCh38
- UniGene ID
- Hs.374477
- Species
- Homo sapiens
- Gene Symbol
- FAM138A
- Entrez Gene ID
- 645520
- Gene Name
- family with sequence similarity 138 member A
- Gene Aliases
- F379, FAM138F
- Chromosome Location
- Chr. 1: 34611 - 36081 on Build GRCh38
- UniGene ID
- Hs.569137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026818.1 |
|
2 |
433 |
|
GenBank mRNA |
AY341950.1 |
- |
2 |
429 |
|
- Gene Symbol
- FAM138C
- Entrez Gene ID
- 654835
- Gene Name
- family with sequence similarity 138 member C
- Gene Aliases
- F379
- Chromosome Location
- Chr. 9: 34394 - 35864 on Build GRCh38
- UniGene ID
- Hs.569137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026822.1 |
|
2 |
433 |
|
- Gene Symbol
- FAM138E
- Entrez Gene ID
- 100124412
- Gene Name
- family with sequence similarity 138 member E
- Gene Aliases
- F379
- Chromosome Location
- Chr. 15: 101954885 - 101956355 on Build GRCh38
- UniGene ID
- Hs.569137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026819.1 |
|
2 |
433 |
|
- Gene Symbol
- FAM138F
- Entrez Gene ID
- 641702
- Gene Name
- family with sequence similarity 138 member F
- Gene Aliases
- F379
- Chromosome Location
- Chr. 19: 76220 - 77690 on Build GRCh38
- UniGene ID
- Hs.569137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026820.1 |
|
2 |
433 |
|
GenBank mRNA |
AY341952.1 |
- |
2 |
429 |
|
BC137207.1 |
- |
2 |
265 |
|
- Gene Symbol
- FNBP1
- Entrez Gene ID
- 23048
- Gene Name
- formin binding protein 1
- Gene Aliases
- FBP17
- Chromosome Location
- Chr. 9: 129887187 - 130053846 on Build GRCh38
- UniGene ID
- Hs.189409
- Species
- Homo sapiens
- Gene Symbol
- GTF2H2B
- Entrez Gene ID
- 653238
- Gene Name
- general transcription factor IIH subunit 2B (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 5: 70415370 - 70450362 on Build GRCh38
- UniGene ID
- Hs.607501
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537982.1 |
- |
7 |
1791 |
|
- Gene Symbol
- IKZF3
- Entrez Gene ID
- 22806
- Gene Name
- IKAROS family zinc finger 3
- Gene Aliases
- AIO, AIOLOS, ZNFN1A3
- Chromosome Location
- Chr. 17: 39757715 - 39864188 on Build GRCh38
- UniGene ID
- Hs.371680
- Species
- Homo sapiens
- Gene Symbol
- IQSEC2
- Entrez Gene ID
- 23096
- Gene Name
- IQ motif and Sec7 domain 2
- Gene Aliases
- BRAG1, MRX1, MRX78
- Chromosome Location
- Chr. X: 53225784 - 53321350 on Build GRCh38
- UniGene ID
- Hs.496138
- Species
- Homo sapiens
- Gene Symbol
- JAK3
- Entrez Gene ID
- 3718
- Gene Name
- Janus kinase 3
- Gene Aliases
- JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
- Chromosome Location
- Chr. 19: 17824782 - 17848071 on Build GRCh38
- UniGene ID
- Hs.515247
- Species
- Homo sapiens
- Gene Symbol
- LOC100130872
- Entrez Gene ID
- 100130872
- Gene Name
- uncharacterized LOC100130872
- Gene Aliases
- Chromosome Location
- Chr. 4: 1195783 - 1208962 on Build GRCh38
- UniGene ID
- Hs.302963
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024569.1 |
|
4 |
2954 |
|
- Gene Symbol
- MAT2B
- Entrez Gene ID
- 27430
- Gene Name
- methionine adenosyltransferase 2B
- Gene Aliases
- MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR
- Chromosome Location
- Chr. 5: 163503064 - 163519353 on Build GRCh38
- UniGene ID
- Hs.54642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096178.1 |
- |
2 |
1369 |
|
- Gene Symbol
- MFSD4B
- Entrez Gene ID
- 91749
- Gene Name
- major facilitator superfamily domain containing 4B
- Gene Aliases
- KIAA1919, NaGLT1
- Chromosome Location
- Chr. 6: 111259279 - 111280005 on Build GRCh38
- UniGene ID
- Hs.400572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB067506.1 |
- |
1 |
3551 |
|
- Gene Symbol
- PAQR8
- Entrez Gene ID
- 85315
- Gene Name
- progestin and adipoQ receptor family member 8
- Gene Aliases
- C6orf33, LMPB1, MPRB
- Chromosome Location
- Chr. 6: 52362128 - 52407777 on Build GRCh38
- UniGene ID
- Hs.239388
- Species
- Homo sapiens
- Gene Symbol
- PARP9
- Entrez Gene ID
- 83666
- Gene Name
- poly(ADP-ribose) polymerase family member 9
- Gene Aliases
- ARTD9, BAL, BAL1, MGC:7868
- Chromosome Location
- Chr. 3: 122527910 - 122564676 on Build GRCh38
- UniGene ID
- Hs.518200
- Species
- Homo sapiens
- Gene Symbol
- SDHAF2
- Entrez Gene ID
- 54949
- Gene Name
- succinate dehydrogenase complex assembly factor 2
- Gene Aliases
- C11orf79, PGL2, SDH5
- Chromosome Location
- Chr. 11: 61430125 - 61446767 on Build GRCh38
- UniGene ID
- Hs.313247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648946.1 |
- |
1 |
3169 |
|
- Gene Symbol
- SLC25A29
- Entrez Gene ID
- 123096
- Gene Name
- solute carrier family 25 member 29
- Gene Aliases
- C14orf69, CACL, ORNT3
- Chromosome Location
- Chr. 14: 100277986 - 100306547 on Build GRCh38
- UniGene ID
- Hs.578109
- Species
- Homo sapiens
- Gene Symbol
- SUSD3
- Entrez Gene ID
- 203328
- Gene Name
- sushi domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 9: 93058688 - 93085138 on Build GRCh38
- UniGene ID
- Hs.88417
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL137623.1 |
- |
|
528 |
|
- Gene Symbol
- TMEM14B
- Entrez Gene ID
- 81853
- Gene Name
- transmembrane protein 14B
- Gene Aliases
- Chromosome Location
- Chr. 6: 10747759 - 10759774 on Build GRCh38
- UniGene ID
- Hs.667371
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI343263.1 |
- |
1 |
429 |
|
BQ045109.1 |
- |
1 |
406 |
|
- Gene Symbol
- UTP14A
- Entrez Gene ID
- 10813
- Gene Name
- UTP14A small subunit processome component
- Gene Aliases
- NYCO16, SDCCAG16, dJ537K23.3
- Chromosome Location
- Chr. X: 129906121 - 129929762 on Build GRCh38
- UniGene ID
- Hs.458598
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094408.1 |
- |
2 |
1520 |
|
- Gene Symbol
- ZNF329
- Entrez Gene ID
- 79673
- Gene Name
- zinc finger protein 329
- Gene Aliases
- Chromosome Location
- Chr. 19: 58126328 - 58155169 on Build GRCh38
- UniGene ID
- Hs.458377
- Species
- Homo sapiens
View Full Product Details