Gene Transcripts
- Gene Symbol
- POLR2J4
- Entrez Gene ID
- 84820
- Gene Name
- RNA polymerase II subunit J4, pseudogene
- Gene Aliases
- RPB11-phi
- Chromosome Location
- Chr. 7: 43940895 - 44019194 on Build GRCh38
- UniGene ID
- Hs.657028
- Species
- Homo sapiens
- Gene Symbol
- POLR2J
- Entrez Gene ID
- 5439
- Gene Name
- RNA polymerase II subunit J
- Gene Aliases
- POLR2J1, RPB11, RPB11A, RPB11m, hRPB14
- Chromosome Location
- Chr. 7: 102473100 - 102478999 on Build GRCh38
- UniGene ID
- Hs.654952
- Species
- Homo sapiens
- Gene Symbol
- POLR2J2
- Entrez Gene ID
- 246721
- Gene Name
- RNA polymerase II subunit J2
- Gene Aliases
- HRPB11B, RPB11b1
- Chromosome Location
- Chr. 7: 102636748 - 102671729 on Build GRCh38
- UniGene ID
- Hs.610255
- Species
- Homo sapiens
- Gene Symbol
- POLR2J3
- Entrez Gene ID
- 548644
- Gene Name
- RNA polymerase II subunit J3
- Gene Aliases
- POLR2J2, RPB11b1, RPB11b2
- Chromosome Location
- Chr. 7: 102537919 - 102572660 on Build GRCh38
- UniGene ID
- Hs.610255
- Species
- Homo sapiens
View Full Product Details