Gene Transcripts

Gene Symbol
Lhpp
Entrez Gene ID
361663
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Gene Aliases
Chromosome Location
Chr. 1: 204617825 - 204708988 on Build Rnor_6.0
UniGene ID
Rn.15275
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009706.1 NP_001009706.1 7 1034
XM_008759912.1 XP_008758134.1 7 2419
GenBank mRNA BC088448.1 - 7 1034

View Full Product Details