Gene Transcripts

Gene Symbol
Ddx19a
Entrez Gene ID
292022
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
Gene Aliases
Ddx19, ZD10B
Chromosome Location
Chr. 19: 43251510 - 43271866 on Build Rnor_6.0
UniGene ID
Rn.228949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005381.2 NP_001005381.2 7 624
XM_008772490.1 XP_008770712.1 410
GenBank mRNA AY303540.1 - 642
BC079094.1 - 7 595 7123989

View Full Product Details