Gene Transcripts

Gene Symbol
Aldh1b1
Entrez Gene ID
298079
Gene Name
aldehyde dehydrogenase 1 family, member B1
Gene Aliases
Chromosome Location
Chr. 5: 61382351 - 61387359 on Build Rnor_6.0
UniGene ID
Rn.12547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011975.1 NP_001011975.1 2 183
GenBank mRNA BC081884.1 - 2 183

View Full Product Details