Gene Transcripts

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
Chromosome Location
Chr. 19: 18442663 - 18522129 on Build GRCh38
UniGene ID
Hs.515260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006532.3 NP_006523.1 12 3274
XM_017027335.1 XP_016882824.1 13 3258
XM_017027336.1 XP_016882825.1 8 3010
XM_017027337.1 XP_016882826.1 7 2450
GenBank mRNA AB208794.1 - 13 3255
AF157562.1 - 1 211
AL136771.1 - 1 244
BC064558.1 - 1 332
BC108256.1 - 1 416

View Full Product Details