Gene Transcripts
- Gene Symbol
- ZNF276
- Entrez Gene ID
- 92822
- Gene Name
- zinc finger protein 276
- Gene Aliases
- CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
- Chromosome Location
- Chr. 16: 89720368 - 89740925 on Build GRCh38
- UniGene ID
- Hs.290154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113525.1 |
NP_001106997.1
|
9, 10 |
1581 |
|
NM_152287.3 |
NP_689500.2
|
9, 10 |
1571 |
|
NR_110122.1 |
|
10 |
1656 |
|
NR_110126.1 |
|
9, 10 |
1539 |
|
NR_110128.1 |
|
9, 10 |
1462 |
|
NR_110129.1 |
|
9, 10 |
1551 |
|
XM_005256324.3 |
XP_005256381.1
|
9, 10 |
1530 |
|
XM_005256328.3 |
XP_005256385.1
|
7, 8 |
1211 |
|
XM_017023890.1 |
XP_016879379.1
|
8, 9 |
1506 |
|
GenBank mRNA |
AK026482.1 |
- |
9, 10 |
1521 |
|
AK056405.1 |
- |
10 |
1607 |
|
AK096762.1 |
- |
2, 3 |
2160 |
|
AK289801.1 |
- |
9, 10 |
1430 |
|
AK295770.1 |
- |
7, 8 |
1243 |
|
AK315793.1 |
- |
6, 7 |
938 |
|
BC032781.1 |
- |
8, 9 |
1240 |
|
BC038839.1 |
- |
1, 2 |
63 |
|
BC105959.1 |
- |
9, 10 |
1364 |
40036181 |
BC107603.1 |
- |
9, 10 |
1396 |
40036180 |
BC110348.1 |
- |
9, 10 |
1296 |
5196957 |
View Full Product Details