Gene Transcripts
- Gene Symbol
- Ahcy
- Entrez Gene ID
- 29443
- Gene Name
- adenosylhomocysteinase
- Gene Aliases
- Chromosome Location
- Chr. 3: 150587833 - 150603057 on Build Rnor_6.0
- UniGene ID
- Rn.5878
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017201.1 |
NP_058897.1
|
4 |
341 |
|
GenBank mRNA |
M15185.1 |
- |
4 |
341 |
|
View Full Product Details