Gene Transcripts

Gene Symbol
Zfp961
Entrez Gene ID
234413
Gene Name
zinc finger protein 961
Gene Aliases
A230105L22Rik, BC065788, mszf37, mszf55-2
Chromosome Location
Chr. 8: 71951066 - 71970333 on Build GRCm38
UniGene ID
Mm.334607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164581.1 NP_001158053.1 4 2649
GenBank mRNA BC049349.1 - 4 1879

View Full Product Details